missing translation for 'onlineSavingsMsg'
Learn More

1,8-Dinitropyrene (90%), TRC

Product Code. 30135494
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135494 25 mg 25mg
30009879 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30135494 Supplier Toronto Research Chemicals Supplier No. TRCD48027025MG
 Restricted Item
Estimated Shipment: 02-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42397-65-9
Molecular Formula C16 H8 N2 O4
Molecular Weight (g/mol) 292.25
Synonym Pyrene, 1,8-dinitro-, 1,8-Dinitropyrene
IUPAC Name 1,8-dinitropyrene
SMILES [O-][N+](=O)c1ccc2ccc3ccc(c4ccc1c2c34)[N+](=O)[O-]

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C16H8N2O4/c19-17(20)13-7-3-9-1-2-10-4-8-14(18(21)22)12-6-5-11(13)15(9)16(10)12/h1-8H
Formula Weight 292.0484
Chemical Name or Material 1,8-Dinitropyrene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.