missing translation for 'onlineSavingsMsg'
Learn More

Trajan Scientific and Medical™ 1/8 in. Graphite Fittings

Product Code. 15268863
Change view
Click to view available options
Quantity:
10/Pk.
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15268863 10/Pk. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15268863 Supplier Trajan Scientific and Medical™ Supplier No. 072609
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

15% Graphite / 85% Vespel ferrules provide leak-free system connections, leading to improved chromatography for non-MS systems

  • Composite of graphite and Vespel
  • Mechanically robust, hard material, long lifetime
  • Forms a strong grip with the capillary column

Specifications

Type Graphite Reducing Ferrule 1/8 in. Fitting 1.2x6.4x4mm
For Use With (Application) Gas Chromatography
Size (Metric) 6.4 x 4 mm
For Use With (Equipment) 1/8 in. Fittings
Material Graphite
Quantity 10/Pk.
Diameter (Metric) Inside 1.2 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.