Learn More
Trajan Scientific and Medical™ 1/8 in. Graphite/Vespel Fittings
Provide leak-free system connections. Trajan Scientific and Medical 1/8 in. Graphite/Vespel Fittings leads to improved chromatography for non-MS systems.
Brand:  Trajan Scientific and Medical™ 0726700
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15228903
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
Description
Description
  • Easy to use
  • Forms reliable seal
  • Reusable
  • Soft material, forms soft grip to the capillary column
  • Low emissions
  • Composite of graphite and Vespel
  • Mechanically robust, hard material, long lifetime
  • Forms a strong grip with the capillary column
Specifications
Specifications
6.4 mm x 7.4 mm
6.4 mm
7.4 mm
2.4 mm
Gas Chromatography
15% Graphite, 85% Vespel
Graphite Vespel Reducing Ferrule 1/8 in. Fitting 2.4mm x 6.4mm x 7.4mm