missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,8-Octanedioic-d12 Acid, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.10g
0.25g
Specifications
Specifications
| Chemical Name or Material | Octanedioic Acid-D12 |
| CAS | 169397-99-3 |
| Molecular Formula | C8 D12 H2 O4 |
| Quantity | 0.1 g |
| Synonym | Octanedioic-2,2,3,3,4,4,5,5,6,6,7,7-d12 acid (9 CI), Suberic-2,2,3,3,4,4,5,5,6,6,7,7-d12 acid, 1,8-Octanedioic-2,2,3,3,4,4,5,5,6,6,7,7-d12 Acid, 1,6-Hexanedicarboxylic Acid-1,1,2,2,3,3,4,4,5,5,6,6-d12, Octanedioic acid-d12, Octanedioic Acid-D12, Suberic acid-d12 |
| InChI Formula | InChI=1 S/C8H14O4/c9-7(10)5-3-1-2-4-6-8(11)12/h1-6H2,(H,9,10)(H,11,12)/i1D2,2D2,3D2,4D2,5D2,6D2 |
| SMILES | [2 H]C([2 H])(C(=O)O)C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C(=O)O |
| IUPAC Name | 2,2,3,3,4,4,5,5,6,6,7,7-dodecadeuteriooctanedioic acid |
| Molecular Weight (g/mol) | 186.2683 |
| Formula Weight | 186.1645 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction