missing translation for 'onlineSavingsMsg'
Learn More

1,9-Decadiyne, 98%, Thermo Scientific™

Product Code. 10581092 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10581092 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10581092 Supplier Thermo Scientific Acros Supplier No. 351870050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 1720-38-3
CAS Min % 97.5
CAS Max % 100.0
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C10H14
Linear Formula HCidentical toC(CH2)6Cidentical toCH
Refractive Index 1.4490 to 1.4530 (20°C, 589nm)
MDL Number MFCD00014926
Quantity 5 g
Synonym 1,9-decadiyne, acmc-1bppd, 1,9-decadiyne 6ci, 7ci, 8ci, 9ci
InChI Key ILVDYAGPHFWNQI-UHFFFAOYSA-N
SMILES C#CCCCCCCC#C
IUPAC Name deca-1,9-diyne
Molecular Weight (g/mol) 134.22
PubChem CID 74400
ChEBI CHEBI:37823
Formula Weight 134.22
Percent Purity ≥97.5%
Physical Form Liquid
Chemical Name or Material 1, 9-Decadiyne, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,9-Decadiyne
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.