missing translation for 'onlineSavingsMsg'
Learn More

1,9-Diiodononane, TRC

Product Code. 30008241
Change view
Click to view available options
Quantity:
500 mg
2.5 g
Unit Size:
2.5g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008241 500 mg 500mg
30003869 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30008241 Supplier Toronto Research Chemicals Supplier No. TRCD679540500MG

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24613-65-8
Molecular Formula C9H18I2
Molecular Weight (g/mol) 380.05
Synonym NSC 9480, Nonamethylene Diiodide
IUPAC Name 1,9-diiodononane
SMILES ICCCCCCCCCI

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C9H18I2/c10-8-6-4-2-1-3-5-7-9-11/h1-9H2
Formula Weight 379.9498
Chemical Name or Material 1,9-Diiodononane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.