missing translation for 'onlineSavingsMsg'
Learn More

1-Acetonylpyridinium chloride, 97%, ACROS Organics™

Product Code. 10262802 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10262802 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10262802 Supplier Thermo Scientific Acros Supplier No. 352370010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 42508-60-1
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C8H10ClNO
MDL Number MFCD00031949
Quantity 1 g
Synonym 1-acetonylpyridinium chloride, n-acetonylpyridinium chloride, 1-2-oxopropyl pyridinium chloride, 1-2-oxopropyl pyridin-1-ium chloride, acmc-209jpw, 1-acetonylpyridiniumchloride, 1-pyridin-1-ium-1-ylpropan-2-one chloride, 1-2-oxopropyl-2h-pyridin-2-ylium chloride
InChI Key JHYYWKUENNZTMD-UHFFFAOYSA-M
SMILES [Cl-].CC(=O)C[N+]1=CC=CC=C1
IUPAC Name 1-(2-oxopropyl)pyridin-1-ium chloride
Molecular Weight (g/mol) 171.62
PubChem CID 2734121
Formula Weight 171.63
Chemical Name or Material 1-Acetonylpyridinium chloride
Show More Show Less
compliance-icons
Product Identifier
  • 1-Acetonylpyridinium chloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.