missing translation for 'onlineSavingsMsg'
Learn More

(1-Acetoxy-2,2,5,5-tetramethyl-Delta-3-pyrroline-3-methyl) Methanethiosulfonate, TRC

Product Code. 30135683
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135683 50 mg 50mg
30137799 10 mg 10mg
30008226 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135683 Supplier Toronto Research Chemicals Supplier No. TRCA16790050MG
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 392718-69-3
Molecular Formula C12H21NO4S2
Molecular Weight (g/mol) 307.43
Synonym (1-Acetoxy-2,2,5,5-Tetramethyl-D-3-Pyrroline-3-Methyl) Methanesulfonate
IUPAC Name [2,2,5,5-tetramethyl-3-(methylsulfonothioyloxymethyl)pyrrol-1-yl] acetate
SMILES CC(=O)ON1C(C)(C)C=C(COS(=O)(=S)C)C1(C)C

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C12H21NO4S2/c1-9(14)17-13-11(2,3)7-10(12(13,4)5)8-16-19(6,15)18/h7H,8H2,1-6H3
Formula Weight 307.091
Percent Purity >95%
Chemical Name or Material (1-Acetoxy-2,2,5,5-tetramethyl-delta-3-pyrroline-3-methyl) Methanethiosulfonate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.