missing translation for 'onlineSavingsMsg'
Learn More

1-Acetyl-4-cyanopiperidine, TRC

Product Code. 30318877
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30318877 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30318877 Supplier Toronto Research Chemicals Supplier No. TRCA172090500MG
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Acetyl-4-cyanopiperidine
CAS 25503-91-7
Molecular Formula C8 H12 N2 O
Quantity 500 mg
InChI Formula InChI=1S/C8H12N2O/c1-7(11)10-4-2-8(6-9)3-5-10/h8H,2-5H2,1H3
SMILES CC(=O)N1CCC(CC1)C#N
IUPAC Name 1-acetylpiperidine-4-carbonitrile
Molecular Weight (g/mol) 152.19
Formula Weight 152.095
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.