Learn More
1-Amino-imidazolidine-2,4-dione, TRC
Product Code. 30006304
Change view
Click to view available options
Quantity:
250 mg
500 mg
2.5 g
Unit Size:
2.5g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30006304 250 mg 250mg
30005827 500 mg 500mg
30005757 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30006304 Supplier Toronto Research Chemicals Supplier No. TRCA611738250MG
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 6301-02-6
Molecular Formula C3 H5 N3 O2
Molecular Weight (g/mol) 115.09
Synonym 1-Aminohydantoin
IUPAC Name 1-aminoimidazolidine-2,4-dione
SMILES NN1CC(=O)NC1=O
Specifications
Analyte or Component Names 1-Aminohydantoin
Shipping Condition Room Temperature
InChI Formula InChI=1S/C3H5N3O2/c4-6-1-2(7)5-3(6)8/h1,4H2,(H,5,7,8)
Solution Type Neat
Industry Type Pharmaceutical
Quantity 250 mg
Formula Weight 115.0382
Percent Purity >95%
Chemical Name or Material 1-Aminohydantoin
Grade HPLC