missing translation for 'onlineSavingsMsg'
Learn More

1-Aminoindane Hydrochloride, TRC

Product Code. 17962985
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17962985 5 g 5g
17912015 1 g 1g
17934035 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17962985 Supplier Toronto Research Chemicals Supplier No. TRCA6122605G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70146-15-5
Molecular Formula C9 H11 N . Cl H
Molecular Weight (g/mol) 169.65
Synonym 1H-Inden-1-amine, 2,3-dihydro-, hydrochloride (1:1), 1-Indanamine, hydrochloride (6CI,7CI), 1H-Inden-1-amine, 2,3-dihydro-, hydrochloride (9CI), 1-Aminoindan hydrochloride, 1-Aminoindane hydrochloride, NSC 186227, Indan-1-amine Hydrochloride
IUPAC Name 2,3-dihydro-1H-inden-1-amine;hydrochloride
SMILES Cl.NC1CCc2ccccc12

Specifications

Quantity 5 g
InChI Formula InChI=1S/C9H11N.ClH/c10-9-6-5-7-3-1-2-4-8(7)9;/h1-4,9H,5-6,10H2;1H
Formula Weight 169.0658
Chemical Name or Material 1-Aminoindane Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.