missing translation for 'onlineSavingsMsg'
Learn More

1-(Azidomethyl)pyrene, TRC

Product Code. 30334160
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
500mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30334160 50 mg 50mg
30343201 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30334160 Supplier Toronto Research Chemicals Supplier No. TRCA91994550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-(Azidomethyl)pyrene
CAS 1006061-57-9
Molecular Formula C17H11N3
Quantity 50 mg
InChI Formula InChI=1S/C17H11N3/c18-20-19-10-14-7-6-13-5-4-11-2-1-3-12-8-9-15(14)17(13)16(11)12/h1-9H,10H2
SMILES [N-]=[N+]=NCc1ccc2ccc3cccc4ccc1c2c34
IUPAC Name 1-(azidomethyl)pyrene
Molecular Weight (g/mol) 257.29
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.