missing translation for 'onlineSavingsMsg'
Learn More

1-Benzyl-4-phenylamino-4-(hydroxymethyl)piperidine, TRC

Product Code. 30003321
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30003321 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30003321 Supplier Toronto Research Chemicals Supplier No. TRCB28858010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 61086-04-2
Molecular Formula C19 H24 N2 O
Quantity 10 mg
Synonym 4-Anilino-1-benzyl-4-piperidinemethanol, 4-(Phenylamino)-1-(phenylmethyl)-4-piperidinemethanol;
InChI Formula InChI=1S/C19H24N2O/c22-16-19(20-18-9-5-2-6-10-18)11-13-21(14-12-19)15-17-7-3-1-4-8-17/h1-10,20,22H,11-16H2
SMILES OCC1(CCN(Cc2ccccc2)CC1)Nc3ccccc3
IUPAC Name (4-anilino-1-benzylpiperidin-4-yl)methanol
Molecular Weight (g/mol) 296.41
Formula Weight 296.1889
Chemical Name or Material 1-Benzyl-4-phenylamino-4-(hydroxymethyl)piperidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.