missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-4-iodopiperidine, 95%

Product Code. 15407007
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15407007 1 g 1g
15417007 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15407007 Supplier Thermo Scientific Alfa Aesar Supplier No. H34090.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

CAS: 301673-14-3 | C10H18INO2 | 311.163 g/mol

Used in the coupling reaction in Iron-and Cobalt-Catalyzed Arylation of Azetidines, Pyrrolidines, and Piperidines with Grignard Reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in the coupling reaction in Iron-and Cobalt-Catalyzed Arylation of Azetidines, Pyrrolidines, and Piperidines with Grignard Reagents.

Solubility
Slightly soluble in water. Soluble in methanol.

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 301673-14-3
Molecular Formula C10H18INO2
Molecular Weight (g/mol) 311.163
MDL Number MFCD04115041
InChI Key YFWQFKUQVJNPKP-UHFFFAOYSA-N
Synonym n-boc-4-iodopiperidine, 1-boc-4-iodopiperidine, n-boc-4-iodo-piperidine, 1-tert-butoxycarbonyl-4-iodopiperidine, tert-butyl 4-iodotetrahydro-1 2h-pyridinecarboxylate, 1-n-boc-4-iodopiperidine, 4-iodo-piperidine-1-carboxylic acid tert-butyl ester, tert-butyl 4-iodopiperidinecarboxylate, tert-butyl 4-iodo-1-piperidinecarboxylate
PubChem CID 10892302
IUPAC Name tert-butyl 4-iodopiperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(CC1)I

Specifications

Quantity 1 g
Sensitivity Light sensitive
Solubility Information Slightly soluble in water. Soluble in methanol.
Formula Weight 311.16
Percent Purity 95%
Chemical Name or Material 1-Boc-4-iodopiperidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.