missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2,5-dichloro-3-fluorobenzene, 97%, Thermo Scientific Chemicals

Product Code. 11329355
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11329355 25 g 25g
11309355 1 g 1g
11319355 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11329355 Supplier Thermo Scientific Alfa Aesar Supplier No. L16713.14
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 202865-57-4
Molecular Formula C6H2BrCl2F
Molecular Weight (g/mol) 243.88
MDL Number MFCD00142585
InChI Key CAYJMDVKWMVOLG-UHFFFAOYSA-N
Synonym 2,5-dichloro-3-fluorobromobenzene, 3-bromo-2,5-dichloro-1-fluorobenzene, pubchem3211, acmc-1cm10, #, 2-bromo-6-fluoro-1,4-phenylene dichloride, benzene, 1-bromo-2,5-dichloro-3-fluoro, 1-bromanyl-2,5-bis chloranyl-3-fluoranyl-benzene
PubChem CID 618513
IUPAC Name 1-bromo-2,5-dichloro-3-fluorobenzene
SMILES FC1=CC(Cl)=CC(Br)=C1Cl

Spécification

Refractive Index 1.5757
Quantity 25 g
Formula Weight 243.89
Percent Purity 97%
Chemical Name or Material 1-Bromo-2,5-dichloro-3-fluorobenzene

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.