Learn More
1-Bromo-2-isopropoxybenzene, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
1-Bromo-2-isopropoxybenzene is used as an intermediate in organic syntheses.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Bromo-2-isopropoxybenzene is used as an intermediate in organic syntheses.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.
1-Bromo-2-isopropoxybenzene is used as an intermediate in organic syntheses.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.
Specifications
Density | 1.34 |
Boiling Point | 73°C (2 mmHg) |
Odor | Aromatic |
Quantity | 25 g |
Solubility Information | Slightly soluble in water. |
Formula Weight | 215.09 |
Percent Purity | 97% |
Chemical Name or Material | 1-Bromo-2-isopropoxybenzene |