missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromo-3,4-difluorobenzene-d3, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | 1-Bromo-3,4-difluorobenzene-D3 |
| CAS | 1219799-14-0 |
| Molecular Formula | C6 D3 Br F2 |
| Quantity | 0.25 g |
| Synonym | 1-Bromo-3,4-difluorobenzene-d3, 1-Bromo-3,4-difluorobenzene-2,5,6-d3 |
| InChI Formula | InChI=1 S/C6H3BrF2/c7-4-1-2-5(8)6(9)3-4/h1-3 H/i1D,2 D,3 D |
| SMILES | [2 H]c1c([2 H])c(Br)c([2 H])c(F)c1F |
| IUPAC Name | 1-bromo-2,3,6-trideuterio-4,5-difluorobenzene |
| Molecular Weight (g/mol) | 196.0073 |
| Formula Weight | 194.9574 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction