missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3,5-dichlorobenzene, 97%

Product Code. 11407176
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11407176 5 g 5g
11417176 25 g 25g
11427176 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11407176 Supplier Thermo Scientific Alfa Aesar Supplier No. B25492.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 19752-55-7 | C6H3BrCl2 | 225.894 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19752-55-7
Molecular Formula C6H3BrCl2
Molecular Weight (g/mol) 225.894
MDL Number MFCD00000584
InChI Key DZHFFMWJXJBBRG-UHFFFAOYSA-N
Synonym 3,5-dichlorobromobenzene, benzene, 1-bromo-3,5-dichloro, 1-bromo-3,5-dichloro-benzene, 3,5-dichloro bromobenzene, 3,5-dichloro-1-bromobenzene, attercop-chm at113311, maybridge1_000881, pubchem3612, acmc-209f1a
PubChem CID 29766
IUPAC Name 1-bromo-3,5-dichlorobenzene
SMILES C1=C(C=C(C=C1Cl)Br)Cl

Specifications

Melting Point 73°C to 77°C
Boiling Point 232°C
Quantity 5 g
Beilstein 1931524
Formula Weight 225.91
Percent Purity 97%
Chemical Name or Material 1-Bromo-3,5-dichlorobenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.