missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromo-4-chlorobutane-d8, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | 1-Bromo-4-chlorobutane-D8 |
| CAS | 1219803-72-1 |
| Molecular Formula | C4 D8 Br Cl |
| Quantity | 0.5 g |
| Synonym | Butane-1,1,2,2,3,3,4,4-d8, 1-bromo-4-chloro- (ACI), 1-Bromo-4-chlorobutane-1,1,2,2,3,3,4,4-d8 (ACI), 1-Bromo-4-chlorobutane-d8, 1-Bromo-4-chlorobutane-D8 |
| InChI Formula | InChI=1 S/C4H8BrCl/c5-3-1-2-4-6/h1-4H2/i1D2,2D2,3D2,4D2 |
| SMILES | [2 H]C([2 H])(Cl)C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])Br |
| IUPAC Name | 1-bromo-4-chloro-1,1,2,2,3,3,4,4-octadeuteriobutane |
| Molecular Weight (g/mol) | 179.5126 |
| Formula Weight | 178 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction