missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-phenylbutane, TRC

Product Code. 30345839
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30345839 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30345839 Supplier Toronto Research Chemicals Supplier No. TRCB695775500MG
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Bromo-4-phenylbutane
CAS 13633-25-5
Molecular Formula C10 H13 Br
Quantity 500 mg
InChI Formula InChI=1S/C10H13Br/c11-9-5-4-8-10-6-2-1-3-7-10/h1-3,6-7H,4-5,8-9H2
SMILES BrCCCCc1ccccc1
IUPAC Name 4-bromobutylbenzene
Molecular Weight (g/mol) 213.114
Formula Weight 212.02
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.