Learn More
1-Bromo-5-chloropentane, 98%
Product Code. 11434467
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11434467 50 g 50g
11424467 10 g 10g
11444467 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11434467 Supplier Thermo Scientific Alfa Aesar Supplier No. B20479.18
 Restricted Item
Estimated Shipment: 29-12-2026
Add to basket
missing translation for 'validMainframeMsg'
CAS: 54512-75-3 | C5H10BrCl | 185.49 g/mol
1-Bromo-5-chloropentane, is used as an primary and secondary intermediate. It is also used as a intermediate in organic syntheses.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Bromo-5-chloropentane, is used as an primary and secondary intermediate. It is also used as a intermediate in organic syntheses.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under normal temperatures and pressures. Keep away from strong oxidizing agents, strong acids, strong bases.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 54512-75-3
Molecular Formula C5H10BrCl
Molecular Weight (g/mol) 185.49
MDL Number MFCD00001016
InChI Key PHHNNDKXQVKJEP-UHFFFAOYSA-N
Synonym pentane, 1-bromo-5-chloro, 5-chloropentyl bromide, 5-bromopentyl chloride, 1-bromo-5-chloro-pentane, 1-brom-5-chlorpentan, 5-chlorobromopentane, bromo-5-chloropentane, pubchem15271, bromo-5-chloro-pentane, 5-bromo-1-chloropentane
PubChem CID 96070
IUPAC Name 1-bromo-5-chloropentane
SMILES ClCCCCCBr
Specifications
Density 1.408
Boiling Point 210°C to 212°C
Flash Point 95°C (203°F)
Refractive Index 1.4836
Quantity 50 g
Beilstein 1732341
Solubility Information Insoluble in water.
Formula Weight 185.5
Percent Purity 98%
Chemical Name or Material 1-Bromo-5-chloropentane