missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromobutane-d9, CDN
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Specifications
Specifications
| Chemical Name or Material | 1-Bromobutane-d9 |
| CAS | 98195-36-9 |
| Molecular Formula | C4D9Br |
| Quantity | 1 g |
| Synonym | 1-Bromobutane-d9, 1-Butyl Bromide-d9, Butyl Bromide-d9, n-Butyl Bromide-d9, 4-Bromobutane-1,1,1-d9; |
| InChI Formula | InChI=1 S/C4H9Br/c1-2-3-4-5/h2-4H2,1H3/i1D3,2D2,3D2,4D2 |
| SMILES | BrC([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 1-bromobutane-1,1,2,2,3,3,4,4,4-d9 |
| Molecular Weight (g/mol) | 146.07 |
| Formula Weight | 145.05 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction