missing translation for 'onlineSavingsMsg'
Learn More

1-(Bromomethyl)-4-(trifluoromethoxy)benzene, 97%, Maybridge

Product Code. 10001724
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10001724 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10001724 Supplier Thermo Scientific Maybridge Supplier No. SB01806DA

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 50824-05-0
CAS Min % 97.0
Density 1.5800g/mL
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C8H6BrF3O
Linear Formula CF3OC6H4CH2Br
MDL Number MFCD00061238
Quantity 1 g
Synonym 4-trifluoromethoxy benzyl bromide, 1-bromomethyl-4-trifluoromethoxy benzene, 4-trifluoromethoxybenzyl bromide, 4-trifluoromethoxy benzylbromide, p-trifluoromethoxy benzyl bromide, alpha-bromo-4-trifluoromethoxy toluene, benzene, 1-bromomethyl-4-trifluoromethoxy, 4-trifluoromethoxy-benzyl bromide, 4-bromomethyl phenoxy trifluoromethane, 1-bromomethyl-4-trifluoromethoxy-benzene
InChI Key JDNPUJCKXLOHOW-UHFFFAOYSA-N
SMILES C1=CC(=CC=C1CBr)OC(F)(F)F
IUPAC Name 1-(bromomethyl)-4-(trifluoromethoxy)benzene
Molecular Weight (g/mol) 255.034
PubChem CID 142785
Formula Weight 255.03
Percent Purity 97%
Chemical Name or Material 1-(Bromomethyl)-4-(trifluoromethoxy)benzene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.