missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromopentane-1,1-d2, CDN
Click to view available options
Quantity:
0.5 g
Unit Size:
0.5g
Specifications
Specifications
| Chemical Name or Material | 1-Bromopentane-1,1-D2 |
| CAS | 77734-75-9 |
| Molecular Formula | C5 D2 H9 Br |
| Quantity | 0.5 g |
| Synonym | Pentane-1,1-d2, 1-bromo- (9 CI, ACI), 1-Bromopentane-1,1-d2 (ACI), 1-Bromo-1,1-dideuteriopentane, 1-Bromopentane-d2, 1-Bromopentane-1,1-D2 |
| InChI Formula | InChI=1 S/C5H11Br/c1-2-3-4-5-6/h2-5H2,1H3/i5D2 |
| SMILES | [2 H]C([2 H])(Br)CCCC |
| IUPAC Name | 1-bromo-1,1-dideuteriopentane |
| Molecular Weight (g/mol) | 153.0572 |
| Formula Weight | 152.017 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction