missing translation for 'onlineSavingsMsg'
Learn More

1-Bromoperfluorohexane, 98%

Product Code. 15458999
Click to view available options
Quantity:
5 g
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 15458999

Brand: Thermo Scientific Alfa Aesar L14125.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

CAS: 335-56-8 | C6BrF13 | 398.949 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 335-56-8
Density 1.88
Boiling Point 100°C to 101°C
Flash Point None
Molecular Formula C6BrF13
Linear Formula CF3(CF2)5Br
Refractive Index 1.2995
MDL Number MFCD00042349
Quantity 5 g
Beilstein 1714896
Synonym 1-bromoperfluorohexane, perfluorohexyl bromide, perfluorohexylbromide, tridecafluorohexyl bromide, 1-bromo-tridecafluorohexane, bromoperfluorohexane, acmc-1cmhf, 1-bromotridecafluorohexane, tridecafluoro-1-bromohexane
InChI Key JTYRBFORUCBNHJ-UHFFFAOYSA-N
SMILES C(C(C(C(F)(F)Br)(F)F)(F)F)(C(C(F)(F)F)(F)F)(F)F
IUPAC Name 1-bromo-1,1,2,2,3,3,4,4,5,5,6,6,6-tridecafluorohexane
Molecular Weight (g/mol) 398.949
PubChem CID 92755
Formula Weight 398.96
Percent Purity 98%
Chemical Name or Material 1-Bromoperfluorohexane
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.