missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-3-fluoro-2-propanol, 95%

Product Code. 15459059
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15459059 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15459059 Supplier Thermo Scientific Alfa Aesar Supplier No. L17286.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 453-11-2 | C3H6ClFO | 112.528 g/mol

It is a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a pharmaceutical intermediate.

Notes
Store in cool dry conditions in well sealed containers. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Specifications

CAS 453-11-2
Density 1.3
Boiling Point 153°C to 156°C
Molecular Formula C3H6ClFO
Refractive Index 1.428
MDL Number MFCD00045284
Quantity 5 g
UN Number UN1992
Beilstein 1732501
Synonym 1-chloro-3-fluoroisopropanol, 1-chloro-3-fluoro-2-propanol, 2-propanol, 1-chloro-3-fluoro, wln: g1yq1f, sgqdlhbrbrwup@, glycerolchlorofluorohydrin, 1-chloro-3-fluoro-propan-2-ol, 5-bromo-2,4-difluorobenzenamine, 1-fluoro-3-chloro isopropanol
InChI Key ZNKJYZHZWALQNX-UHFFFAOYSA-N
SMILES C(C(CCl)O)F
IUPAC Name 1-chloro-3-fluoropropan-2-ol
Molecular Weight (g/mol) 112.528
PubChem CID 92107
Formula Weight 112.53
Percent Purity 95%
Chemical Name or Material 1-Chloro-3-fluoro-2-propanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.