missing translation for 'onlineSavingsMsg'
Learn More

1-Chlorohexadecane, 97%

Product Code. 11461017
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11461017 500 mL 500mL
11451017 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11461017 Supplier Thermo Scientific Alfa Aesar Supplier No. B23406.AP
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4860-03-1 | C9H7F3O3 | 220.15 g/mol

1-Chlorohexadecane is used as solvent , surfactants, pharmaceuticals, antibacterial spray.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chlorohexadecane is used as solvent , surfactants, pharmaceuticals, antibacterial spray.

Solubility
Soluble in ether. Insoluble in water.

Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4860-03-1
MDL Number MFCD00000959

Specifications

Melting Point 8°C to 10°C
Density 0.865
Boiling Point 322°C
Flash Point 136°C (276°F)
Molecular Formula C16H33Cl
Linear Formula CH3(CH2)15Cl
Refractive Index 1.449
Quantity 500 mL
Beilstein 1748498
Synonym cetyl chloride, hexadecyl chloride, hexadecane, 1-chloro, n-hexadecyl chloride, chlorohexadecane, unii-wq5zks3ek3, wq5zks3ek3, 1-chloro-hexadecane, 1-chloranylhexadecane, dsstox_cid_7580
Solubility Information Soluble in ether. Insoluble in water.
InChI Key CLWAXFZCVYJLLM-UHFFFAOYSA-N
SMILES CCCCCCCCCCCCCCCCCl
IUPAC Name 1-chlorohexadecane
Molecular Weight (g/mol) 260.89
PubChem CID 20993
Formula Weight 260.89
Percent Purity 97%
Chemical Name or Material 1-Chlorohexadecane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.