missing translation for 'onlineSavingsMsg'
Learn More

1-Chlorooctadecane 10000 μg/mL in Dichloromethane, Dr. Ehrenstorfer

Product Code. 30310879
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30310879 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30310879 Supplier Dr. Ehrenstorfer Supplier No. DREGA09010318DI
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Chlorooctadecane
CAS 3386-33-2
Analyte or Component Names 1-Chlorooctadecane
Shipping Condition Room Temperature
Molecular Formula C18 H37 Cl
Concentration 10000 μg/mL
Certifications/Compliance ISO 17034
Synonym Octadecane, 1-chloro-, 1-Chlorooctadecane, 1-Octadecyl chloride, NSC 5543, Octadecyl chloride, n-Octadecyl chloride
InChI Formula InChI=1S/C18H37Cl/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19/h2-18H2,1H3
SMILES CCCCCCCCCCCCCCCCCCCl
IUPAC Name 1-chlorooctadecane
Solvent or Matrix Dichloromethane
Molecular Weight (g/mol) 288.94
Quantity 1 mL
Formula Weight 288.2584 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.