missing translation for 'onlineSavingsMsg'
Learn More

1-Cyanotetraline, TRC

Product Code. 30134343
Change view
Click to view available options
Quantity:
5 g
50 g
100 g
Unit Size:
100g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134343 100 g 100g
30136125 50 g 50g
30005208 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134343 Supplier Toronto Research Chemicals Supplier No. TRCC987780100G
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56536-96-0
Molecular Formula C11H11N
Molecular Weight (g/mol) 157.21
Synonym α-Cyanotetraline, (1RS)-1,2,3,4-Tetrahydronaphthalene-1-carbonitrile, Tetryzoline Hydrochloride Imp. A (EP), alpha-Cyanotetraline, 1-Naphthalenecarbonitrile, 1,2,3,4-tetrahydro-, 1-Naphthonitrile, 1,2,3,4-tetrahydro- (6CI,7CI), 1,2,3,4-Tetrahydro-1-naphthalenecarbonitrile, 1-Cyano-1,2,3,4-tetrahydronaphthalene, 1-Cyanotetralin
IUPAC Name 1,2,3,4-tetrahydronaphthalene-1-carbonitrile
SMILES N#CC1CCCc2ccccc12

Specifications

Quantity 100 g
InChI Formula InChI=1S/C11H11N/c12-8-10-6-3-5-9-4-1-2-7-11(9)10/h1-2,4,7,10H,3,5-6H2
Formula Weight 157.0891
Chemical Name or Material (1RS)-1,2,3,4-Tetrahydronaphthalene-1-carbonitrile (alpha-Cyanotetraline)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.