missing translation for 'onlineSavingsMsg'
Learn More

1-Cyclohexyl-3-methyl-1,3,5-triazinane-2,4,6-trione, TRC

Product Code. 30179844
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179844 100 mg 100mg
30180226 50 mg 50mg
30179761 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30179844 Supplier Toronto Research Chemicals Supplier No. TRCC572355100MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30243-77-7
Molecular Formula C10 H15 N3 O3
Molecular Weight (g/mol) 225.24
Synonym s-Triazine-2,4,6(1H,3H,5H)-trione, 1-cyclohexyl-3-methyl- (8CI), 1-Cyclohexyl-3-methyl-1,3,5-triazine-2,4,6(1H,3H,5H)-trione
IUPAC Name 1-cyclohexyl-3-methyl-1,3,5-triazinane-2,4,6-trione
SMILES CN1C(=O)NC(=O)N(C2CCCCC2)C1=O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C10H15N3O3/c1-12-8(14)11-9(15)13(10(12)16)7-5-3-2-4-6-7/h7H,2-6H2,1H3,(H,11,14,15)
Formula Weight 225.1113
Chemical Name or Material 1-Cyclohexyl-3-methyl-1,3,5-triazine-2,4,6(1H,3H,5H)-trione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.