missing translation for 'onlineSavingsMsg'
Learn More

1-Dimethylaminomethyl-2-naphthol, 98%, Thermo Scientific™

Product Code. 11345459
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11345459 1 g 1g
11335459 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11345459 Supplier Thermo Scientific Alfa Aesar Supplier No. H50430.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5419-02-3
MDL Number MFCD00086150

Specifications

Molecular Formula C16H8N4Na2O11S2
Quantity 1 g
Synonym nitrazine yellow, 2,7-naphthalenedisulfonic acid, 3-2-2,4-dinitrophenyl diazenyl-4-hydroxy-, sodium salt 1:2, disodium 3-e-2-2,4-dinitrophenyl diazen-1-yl-4-hydroxynaphthalene-2,7-disulfonate
InChI Key XBLIFEQTVVSTIM-QIKYXUGXSA-L
SMILES C1=CC(=C(C=C1[N+](=O)[O-])[N+](=O)[O-])NN=C2C(=CC3=C(C2=O)C=CC(=C3)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]
IUPAC Name disodium;3-[(2,4-dinitrophenyl)hydrazinylidene]-4-oxonaphthalene-2,7-disulfonate
Molecular Weight (g/mol) 542.36
PubChem CID 44134855
Formula Weight 201.27
Percent Purity 98%
Chemical Name or Material 1-Dimethylaminomethyl-2-naphthol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.