missing translation for 'onlineSavingsMsg'
Learn More

1-Dodecanol Acetate, TRC

Product Code. 30138447
Change view
Click to view available options
Quantity:
25 g
50 g
100 g
Unit Size:
100g
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138447 100 g 100g
30132976 50 g 50g
30010593 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138447 Supplier Toronto Research Chemicals Supplier No. TRCD525390100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-66-3
Molecular Formula C14H28O2
Molecular Weight (g/mol) 228.37
Synonym Dodecyl Alcohol Acetate, Dodecyl Ester Acetic Acid, 1-Dodecyl Acetate, Dodecan-1-yl Acetate, Dodecanol Acetate, Dodecanyl Acetate, Dodecyl Acetate, Lauryl Acetate, NSC 67366, Okimelanolure, n-Dodecyl Acetate
IUPAC Name dodecyl acetate
SMILES CCCCCCCCCCCCOC(=O)C

Specifications

Quantity 100 g
InChI Formula InChI=1S/C14H28O2/c1-3-4-5-6-7-8-9-10-11-12-13-16-14(2)15/h3-13H2,1-2H3
Formula Weight 228.2089
Chemical Name or Material 1-Dodecanol Acetate
Product Title
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.