missing translation for 'onlineSavingsMsg'
Learn More

(1-Ethoxycarbonylcyclopropyl)triphenylphosphonium tetrafluoroborate, 98%

Product Code. 11473857
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11473857 25 g 25g
11453857 1 g 1g
11463857 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11473857 Supplier Thermo Scientific Alfa Aesar Supplier No. A18305.14
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 52186-89-7 | C24H24BF4O2P | 462.23 g/mol

(1-Ethoxycarbonylcyclopropyl)triphenylphosphonium tetrafluoroborate is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(1-Ethoxycarbonylcyclopropyl)triphenylphosphonium tetrafluoroborate is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52186-89-7
Molecular Formula C24H24BF4O2P
Molecular Weight (g/mol) 462.23
MDL Number MFCD00051879
InChI Key RGJYRMUGSAFITK-UHFFFAOYSA-N
Synonym 1-carbethoxycyclopropyltriphenylphosphonium tetrafluoroborate, 1-ethoxycarbonyl cyclopropyl triphenylphosphonium tetrafluoroborate, 1-ethoxycarbonylcyclopropyl triphenylphosphonium tetrafluoroborate, 1-ethoxycarbonyl cyclopropyl tris phenyl phosphonium tetrafluoroborate, 1-ethoxycarbonyl cyclopropyl triphenylphosphanium tetrafluoroborate, acmc-20aowe, 1-carboethoxycyclopropyl triphenylphosphonium tetrafluoroborate, 1-carbethoxycyclopropyl tris phenyl phosphonium tetrafluoroborate, 1-ethoxycarbonylcyclopropyl-triphenyl-phosphanium tetrafluoroborate, 1-ethoxycarbonylcyclopropyl-triphenyl-phosphonium tetrafluoroborate
PubChem CID 11016017
SMILES F[B-](F)(F)F.CCOC(=O)C1(CC1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 173°C to 179°C
Quantity 25 g
Beilstein 4650673
Solubility Information Slightly soluble in water.
IUPAC Name (1-ethoxycarbonylcyclopropyl)-triphenylphosphanium;tetrafluoroborate
Formula Weight 462.23
Percent Purity 98%
Chemical Name or Material (1-Ethoxycarbonylcyclopropyl)triphenylphosphonium tetrafluoroborate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.