missing translation for 'onlineSavingsMsg'
Learn More

1-Ethyl-4-[(p-tolyl)ethynyl]benzene, 99+%, Thermo Scientific Chemicals

Product Code. 11387069
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11387069 1 g 1g
11397069 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11387069 Supplier Thermo Scientific Alfa Aesar Supplier No. H26882.03
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22692-80-4
Molecular Formula C17H16
Molecular Weight (g/mol) 220.315
MDL Number MFCD04038793
InChI Key LFNSILZUJYWPJS-UHFFFAOYSA-N
Synonym 1-4-ethylphenyl-2-4-methylphenyl acetylene, 1-ethyl-4-p-tolyl ethynyl benzene, 1-ethyl-4-2-4-methylphenyl ethynyl benzene, 1-4-ethyl-4-p-tolyl ethylnyl benzene, acmc-20apau, 1-ethyl-4-p-tolylethynyl benzene, 1-4-ethylphenyl-2-p-tolyl acetylene, 1-ethyl-4-4-methylphenyl ethynyl benzene, benzene,1-ethyl-4-2-4-methylphenyl ethynyl, 1-ethyl-4-2-4-methylphenyl-1-ethynyl benzene
PubChem CID 11413368
IUPAC Name 1-ethyl-4-[2-(4-methylphenyl)ethynyl]benzene
SMILES CCC1=CC=C(C=C1)C#CC2=CC=C(C=C2)C

Specifications

Melting Point 71°C to 74°C
Boiling Point 135°C to 136°C (1 mmHg)
Quantity 1 g
Formula Weight 220.31
Percent Purity ≥99%
Chemical Name or Material 1-Ethyl-4-[(p-tolyl)ethynyl]benzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.