missing translation for 'onlineSavingsMsg'
Learn More

1-Fluoro-3,5-dimethoxybenzene, 98%, Thermo Scientific™

Product Code. 10756265 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10756265 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10756265 Supplier Thermo Scientific Acros Supplier No. 399410050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 52189-63-6
Density 1.1710g/mL
Boiling Point 86°C to 88°C (7.0 mmHg)
Flash Point 91°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Molecular Formula C8H9FO2
Linear Formula FC6H3(OCH3)2
MDL Number MFCD00012445
Quantity 5 g
Specific Gravity 1.171
Synonym 1,3-dimethoxy-5-fluorobenzene, benzene, 1-fluoro-3,5-dimethoxy, 3,5-dimethoxyfluorobenzene, pubchem3049, acmc-209wz0, 3,5-dimethoxy fluorobenzene, ksc595k6b, 3,5-dimethoxy-1-fluorobenzene, 1-fluoro-3,5-dimethoxy benzene, benzene,1-fluoro-3,5-dimethoxy
InChI Key IWFKMNAEFPEIOY-UHFFFAOYSA-N
SMILES COC1=CC(OC)=CC(F)=C1
IUPAC Name 1-fluoro-3,5-dimethoxybenzene
Molecular Weight (g/mol) 156.16
PubChem CID 2774257
Formula Weight 156.16
Percent Purity 98%
Chemical Name or Material 1-Fluoro-3, 5-dimethoxybenzene
Show More Show Less
compliance-icons
Product Identifier
  • 1-Fluoro-3,5-dimethoxybenzene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.