missing translation for 'onlineSavingsMsg'
Learn More

1-Formyl-4-methylpiperazine, 98%

Product Code. 11334286
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11334286 5 g 5g
11344286 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11334286 Supplier Thermo Scientific Alfa Aesar Supplier No. L19358.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 7556-55-0 | C6H12N2O | 128.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7556-55-0
Molecular Formula C6H12N2O
Molecular Weight (g/mol) 128.18
MDL Number MFCD00085930
InChI Key JQTMGOLZSBTZMS-UHFFFAOYSA-N
Synonym 1-formyl-4-methylpiperazine, n-methyl-n'-formylpiperazine, 4-methylpiperazin-1-yl methanone, 4-methyl-1-piperazinyl-methanone, 4-methyl-piperazin-1-yl-methanone, ods
PubChem CID 3269221
ChEBI CHEBI:44477
IUPAC Name 4-methylpiperazine-1-carbaldehyde
SMILES CN1CCN(CC1)C=O

Specifications

Boiling Point 236°C to 238°C
Flash Point >110°C (230°F)
Refractive Index 1.491
Quantity 5 g
Beilstein 113020
Formula Weight 128.17
Percent Purity 98%
Chemical Name or Material 1-Formyl-4-methylpiperazine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.