missing translation for 'onlineSavingsMsg'
Learn More

1-Formylpiperazine, tech. 90

Product Code. 11487806
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11487806 25 g 25g
11497806 100 g 100g
11407816 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11487806 Supplier Thermo Scientific Alfa Aesar Supplier No. A14757.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 7755-92-2 | C5H11N2O | 115.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7755-92-2
Molecular Formula C5H11N2O
Molecular Weight (g/mol) 115.16
MDL Number MFCD00005963
InChI Key MSSDTZLYNMFTKN-UHFFFAOYSA-O
Synonym 1-formylpiperazine, 1-piperazinecarboxaldehyde, 1-piperazinecarbaldehyde, formylpiperazine, n-formylpiperazine, 1-formyl piperazine, piperazinecarbaldehyde, l-formylpiperazine, 4-formylpiperazine, n-formyl piperazine
PubChem CID 82191
IUPAC Name piperazine-1-carbaldehyde
SMILES O=CN1CC[NH2+]CC1

Specifications

Density 1.111
Boiling Point 264°C to 266°C
Flash Point 102°C (216°F)
Refractive Index 1.512
Quantity 25 g
Beilstein 110295
Formula Weight 114.15
Percent Purity ≥90%
Chemical Name or Material 1-Formylpiperazine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.