missing translation for 'onlineSavingsMsg'
Learn More

1-Hexen-3-one (>90%), TRC

Product Code. 30391022
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. Quantity unitSize
30391022 1 g 1g
1 options
This item is not returnable. View return policy

Product Code. 30391022

Brand: Toronto Research Chemicals TRCH2950201G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Hexen-3-one
CAS 1629-60-3
Molecular Formula C6H10O
Quantity 1 g
Synonym Propyl Vinyl Ketone, Vinyl Propyl Ketone
InChI Formula InChI=1S/C6H10O/c1-3-5-6(7)4-2/h4H,2-3,5H2,1H3
SMILES O=C(C=C)CCC
IUPAC Name hex-1-en-3-one
Molecular Weight (g/mol) 98.14
Formula Weight 98.0732
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.