missing translation for 'onlineSavingsMsg'
Learn More

1-Hydroxy-2-(2-pyridinyl) Risedronate, TRC

Product Code. 30136759
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136759 50 mg 50mg
30005313 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30136759 Supplier Toronto Research Chemicals Supplier No. TRCH95281050MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105462-23-5
Molecular Formula C7 H11 N O7 P2
Molecular Weight (g/mol) 283.11
Synonym [1-Hydroxy-2-(pyridin-2-yl)ethane-1,1-diyl]bis(phosphonic Acid), P,P'-[1-Hydroxy-2-(2-pyridinyl)ethylidene]bis-phosphonic acid, [1-Hydroxy-2-(2-pyridinyl)ethylidene]bis-phosphonic acid, NE 58018
IUPAC Name (1-hydroxy-1-phosphono-2-pyridin-2-ylethyl)phosphonic acid
SMILES OC(Cc1ccccn1)(P(=O)(O)O)P(=O)(O)O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C7H11NO7P2/c9-7(16(10,11)12,17(13,14)15)5-6-3-1-2-4-8-6/h1-4,9H,5H2,(H2,10,11,12)(H2,13,14,15)
Formula Weight 283.0011
Chemical Name or Material (1-Hydroxy-2-(pyridin-2-yl)ethylidene)bis(phosphonic Acid) ((1-Hydroxy-2-(pyridin-2-yl)ethane-1,1-diyl)bis(phosphonic Acid))
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.