missing translation for 'onlineSavingsMsg'
Learn More

1-Hydroxy-2-naphthoic acid, 98+%, Thermo Scientific™

Product Code. 11434170
Change view
Click to view available options
Quantity:
250 g
500 g
1000 g
Unit Size:
1000g
250g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11434170 500 g 500g
11425133 250 g 250g
11415133 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11434170 Supplier Thermo Scientific Alfa Aesar Supplier No. A12785.36

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

Solubility
Soluble in alcohol, ethanol, benzene, methanol, and water (partly).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.Stable under recommended storage conditions. Incompatible to strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86-48-6
Molecular Formula C11H8O3
Molecular Weight (g/mol) 188.182
MDL Number MFCD00003960
InChI Key SJJCQDRGABAVBB-UHFFFAOYSA-N
Synonym 1-hydroxy-2-naphthoic acid, 1-naphthol-2-carboxylic acid, 2-carboxy-1-naphthol, 2-naphthalenecarboxylic acid, 1-hydroxy, 1-hydroxy-2-naphthoate, 1-hydroxy-2-naphthalenecarboxylic acid, unii-u8lz3r07l8, 2-naphthoic acid, 1-hydroxy, alpha-hydroxynaphthoic acid, 1-hydroxy-naphthalene-2-carboxylic acid
PubChem CID 6844
ChEBI CHEBI:36108
IUPAC Name 1-hydroxynaphthalene-2-carboxylic acid
SMILES C1=CC=C2C(=C1)C=CC(=C2O)C(=O)O

Specifications

Melting Point ∼200°C (decomposition)
Quantity 500 g
Beilstein 974438
Merck Index 14,4834
Formula Weight 188.18
Percent Purity ≥98%
Chemical Name or Material 1-Hydroxy-2-naphthoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.