missing translation for 'onlineSavingsMsg'
Learn More

1-Hydroxy-2-naphthoic Acid, TRC

Product Code. 30406812
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30406812 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30406812 Supplier Toronto Research Chemicals Supplier No. TRCH95085050G
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Xinafoic acid
CAS 86-48-6
Molecular Formula C11 H8 O3
Quantity 50 g
Synonym 1-Hydroxy-2-naphthoic Acid, 2-Naphthalenecarboxylic acid, 1-hydroxy-, 2-Naphthoic acid, 1-hydroxy- (8CI), 1-Hydroxy-2-naphthalenecarboxylic acid, 1-Hydroxy-2-naphthoic acid, 1-Naphthol-2-carboxylic acid, 2-Carboxy-1-naphthol, NSC 3717, Xinafoic acid, α-Hydroxy-β-naphthoic acid, GR 66288X, Salmeterol related
InChI Formula InChI=1S/C11H8O3/c12-10-8-4-2-1-3-7(8)5-6-9(10)11(13)14/h1-6,12H,(H,13,14)
SMILES C1=CC=C2C(=C1)C=CC(=C2O)C(=O)O
IUPAC Name 1-hydroxynaphthalene-2-carboxylic acid
Molecular Weight (g/mol) 188.18
Formula Weight 188.047
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.