missing translation for 'onlineSavingsMsg'
Learn More

[1-Hydroxy-2-(pyridin-2-yl)ethylidene]bis(phosphonic Acid) ([1-Hydroxy-2-(pyridin-2-yl)ethane-1,1-diyl]bis(phosphonic Acid)), Mikromol™

Product Code. 30052516
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30052516 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30052516 Supplier Mikromol Supplier No. MM0777.020025
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

[1-Hydroxy-2-(pyridin-2-yl)ethylidene]bis(phosphonic Acid) ([1-Hydroxy-2-(pyridin-2-yl)ethane-1,1-diyl]bis(phosphonic Acid))
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (1-Hydroxy-2-(pyridin-2-yl)ethylidene)bis(phosphonic Acid) ((1-Hydroxy-2-(pyridin-2-yl)ethane-1,1-diyl)bis(phosphonic Acid))
CAS 105462-23-5
Analyte or Component Names (1-Hydroxy-2-(pyridin-2-yl)ethylidene)bis(phosphonic Acid) ((1-Hydroxy-2-(pyridin-2-yl)ethane-1,1-diyl)bis(phosphonic Acid))
Shipping Condition Room Temperature
Molecular Formula C7 H11 N O7 P2
Certifications/Compliance ISO 9001
Synonym Risedronate Sodium 2.5 Hydrate Imp. B (EP), Risedronate Imp. B (EP), Risedronate USP Related Compound A, Risedronate USP RC A, Risedronate Sodium 2.5 Hydrate Imp B (Pharmeuropa), [1-Hydroxy-2-(pyridin-2-yl)ethane-1,1-diyl]bis(phosphonic Acid), P,P'-[1-Hydroxy-2-(2-pyridinyl)ethylidene]bis-phosphonic acid, [1-Hydroxy-2-(2-pyridinyl)ethylidene]bis-phosphonic acid, NE 58018, Risedronate Related Compound A, Risedronate Sodium 2.5 Hydrate Impurity B, Risedronate Impurity B
InChI Formula InChI=1S/C7H11NO7P2/c9-7(16(10,11)12,17(13,14)15)5-6-3-1-2-4-8-6/h1-4,9H,5H2,(H2,10,11,12)(H2,13,14,15)
SMILES OC(Cc1ccccn1)(P(=O)(O)O)P(=O)(O)O
IUPAC Name (1-hydroxy-1-phosphono-2-pyridin-2-ylethyl)phosphonic acid
Molecular Weight (g/mol) 283.11
Quantity 25 mg
Formula Weight 283.0011 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.