missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Hydroxypyrene-d9, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | 1-Hydroxypyrene-d9 |
| CAS | 132603-37-3 |
| Molecular Formula | C16 2H9 H O |
| Quantity | 0.05 g |
| Synonym | 1-Pyren-2,3,4,5,6,7,8,9,10-d9-ol, 1-Hydroxypyrene-d9, 1-Hydroxypyrene D9 |
| InChI Formula | InChI=1 S/C16H10O/c17-14-9-7-12-5-4-10-2-1-3-11-6-8-13(14)16(12)15(10)11/h1-9,17 H/i1D,2 D,3 D,4 D,5 D,6 D,7 D,8 D,9 D |
| SMILES | [2 H]c1c([2 H])c2c([2 H])c([2 H])c3c([2 H])c([2 H])c(O)c4c([2 H])c([2 H])c(c1[2 H])c2c34 |
| IUPAC Name | 2,3,4,5,6,7,8,9,10-nonadeuteriopyren-1-ol |
| Molecular Weight (g/mol) | 227.31 |
| Formula Weight | 227.1297 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction