missing translation for 'onlineSavingsMsg'
Learn More

1-Indanone, TRC

Product Code. 30137723
Change view
Click to view available options
Quantity:
10 g
25 g
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137723 25 g 25g
30005037 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30137723 Supplier Toronto Research Chemicals Supplier No. TRCI49990025G
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83-33-0
Molecular Formula C9 H8 O
Molecular Weight (g/mol) 132.16
Synonym 1H-Inden-1-one, 2,3-dihydro-, 1-Indanone (8CI), Indanone (7CI), 2,3-Dihydro-1H-inden-1-one, 1-Indone, 1-Oxobenzocyclopentane, 1H-Indan-1-one, 2,3-Dihydro-1-indenone, NSC 2581, α-Hydrindone, α-Indanone
IUPAC Name 2,3-dihydroinden-1-one
SMILES O=C1CCc2ccccc12

Specifications

Quantity 25 g
InChI Formula InChI=1S/C9H8O/c10-9-6-5-7-3-1-2-4-8(7)9/h1-4H,5-6H2
Formula Weight 132.0575
Chemical Name or Material 1-Indanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.