missing translation for 'onlineSavingsMsg'
Learn More

1-Iodo-4-n-propylbenzene, 97%

Product Code. 11357585
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11357585 5 g 5g
11367585 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11357585 Supplier Thermo Scientific Alfa Aesar Supplier No. L05767.06
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 126261-84-5 | C9H11I | 246.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 126261-84-5
Molecular Formula C9H11I
Molecular Weight (g/mol) 246.09
MDL Number MFCD00051612
InChI Key ZRWCHMFGMKNWEC-UHFFFAOYSA-N
Synonym 1-iodo-4-n-propylbenzene, 1-iodo-4-propyl-benzene, 4-iodopropylbenzene, 4-propyliodobenzene, p-n-propyliodobenzene, 4-iodo-n-propylbenzene, 4-n-propyl iodobenzene, #, benzene,1-iodo-4-propyl, 1-iodanyl-4-propyl-benzene
PubChem CID 612541
IUPAC Name 1-iodo-4-propylbenzene
SMILES CCCC1=CC=C(I)C=C1

Specifications

Boiling Point 240°C to 242°C
Refractive Index 1.579
Quantity 5 g
Sensitivity Light sensitive
Formula Weight 246.09
Percent Purity 97%
Chemical Name or Material 1-Iodo-4-n-propylbenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.