missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-imidazole-4-carbaldehyde, TRC

Product Code. 30481542
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30481542 50 mg 50mg
30465508 500 mg 500mg
30469483 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30481542 Supplier Toronto Research Chemicals Supplier No. TRCM22251350MG
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17289-26-8
Molecular Formula C5H6N2O
Molecular Weight (g/mol) 110.11
IUPAC Name 1-methylimidazole-4-carbaldehyde
SMILES Cn1cnc(C=O)c1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C5H6N2O/c1-7-2-5(3-8)6-4-7/h2-4H,1H3
Formula Weight 110.048
Chemical Name or Material 1-Methyl-1H-imidazole-4-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.