missing translation for 'onlineSavingsMsg'
Learn More

(1-Methyl-1H-pyrazol-5-yl)methylamine, 97%, Maybridge

Product Code. 10751385
Change view
Click to view available options
Amber Glass Bottle; 1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10751385 Amber Glass Bottle; 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10751385 Supplier Thermo Scientific Maybridge Supplier No. CC62313DA

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 863548-52-1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C5H9N3
MDL Number MFCD03419808
Quantity 1 g
Synonym 1-methyl-1h-pyrazol-5-yl methanamine, 1-methyl-1h-pyrazol-5-yl methylamine, 1h-pyrazole-5-methanamine, 1-methyl, 5-aminomethyl-1-methyl-1h-pyrazole, 2-methylpyrazol-3-yl methanamine, 5-aminomethyl-1-methylpyrazole, 1-1-methyl-1h-pyrazol-5-yl methanamine, 1-methyl-1h-pyrazole-5-methanamine, 1-methylpyrazol-5-yl methylamine, c-2-methyl-2h-pyrazol-3-yl-methylamine
InChI Key XNTFQMKXUFFUQO-UHFFFAOYSA-N
SMILES CN1C(=CC=N1)CN
IUPAC Name (2-methylpyrazol-3-yl)methanamine
Molecular Weight (g/mol) 111.148
PubChem CID 7019421
Formula Weight 111.15
Percent Purity 97%
Chemical Name or Material (1-Methyl-1H-pyrazol-5-yl)methylamine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.