missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-pyrazole-4,5-diamine Sulfate, TRC

Product Code. 30472950
Change view
Click to view available options
Quantity:
500 mg
5 g
Unit Size:
500mg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30472950 5 g 5g
30470593 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30472950 Supplier Toronto Research Chemicals Supplier No. TRCM3280355G
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20055-01-0
Molecular Formula C4H10N4O4S
Molecular Weight (g/mol) 210.21
Synonym 4,5-Diamino-1-methylpyrazole Sulfate, 4,5-Diamino-1-methylpyrazole Hydrosulfate
IUPAC Name 2-methylpyrazole-3,4-diamine;sulfuric acid
SMILES Cn1ncc(N)c1N.OS(=O)(=O)O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C4H8N4.H2O4S/c1-8-4(6)3(5)2-7-8;1-5(2,3)4/h2H,5-6H2,1H3;(H2,1,2,3,4)
Formula Weight 210.0423
Chemical Name or Material 1-Methyl-1H-pyrazole-4,5-diamine Sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.