missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-3-n-propylimidazolium iodide, 98%

Product Code. 11372769
Change view
Click to view available options
Quantity:
5 g
50 g
Unit Size:
50g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11372769 5 g 5g
11382769 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11372769 Supplier Thermo Scientific Alfa Aesar Supplier No. H27682.06
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 119171-18-5 | C7H13IN2 | 252.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119171-18-5
Molecular Formula C7H13IN2
Molecular Weight (g/mol) 252.10
MDL Number MFCD08276385
InChI Key IVCMUVGRRDWTDK-UHFFFAOYSA-M
Synonym 1-methyl-3-propylimidazolium iodide, 1-propyl-3-methylimidazolium iodide, 1-methyl-3-n-propylimidazolium iodide, 1-methyl-3-propylimidazol-1-ium iodide, acmc-1c1fw, dsstox_cid_29063, dsstox_rid_83282, dsstox_gsid_49207, ksc491c6t, 1-methyl-3-propylimidazoliumiodide
PubChem CID 12971008
IUPAC Name 1-methyl-3-propylimidazol-1-ium;iodide
SMILES [I-].CCCN1C=C[N+](C)=C1

Specifications

Density 1.54
Boiling Point 301°C to 304°C
Flash Point >110°C (230°F)
Quantity 5 g
Sensitivity Light sensitive
Formula Weight 252.09
Percent Purity 98%
Chemical Name or Material 1-Methyl-3-n-propylimidazolium iodide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.