missing translation for 'onlineSavingsMsg'
Learn More

(1-Methylcyclohexanyl)methyl-4-nitrophenyl Ether, TRC

Product Code. 30478391
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30478391 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30478391 Supplier Toronto Research Chemicals Supplier No. TRCM294625100MG
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (1-Methylcyclohexanyl)methyl-4-nitrophenyl Ether
CAS 85002-76-2
Molecular Formula C14 H19 N O3
Quantity 100 mg
Synonym Benzene, 1-[(1-methylcyclohexyl)methoxy]-4-nitro-, (1-Methylcyclohexanyl)methyl-4-nitrophenyl ether, (1-Methylcyclohexyl)methyl-4-nitrophenyl ether, 1-[(1-Methylcyclohexyl)methoxy]-4-nitrobenzene
InChI Formula InChI=1S/C14H19NO3/c1-14(9-3-2-4-10-14)11-18-13-7-5-12(6-8-13)15(16)17/h5-8H,2-4,9-11H2,1H3
SMILES CC1(COc2ccc(cc2)[N+](=O)[O-])CCCCC1
IUPAC Name 1-[(1-methylcyclohexyl)methoxy]-4-nitrobenzene
Molecular Weight (g/mol) 249.31
Formula Weight 249.1365
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.